Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014235.1 | NP_001014257.1 | 363 | |||
GenBank mRNA | BC088253.1 | - | 363 | 7374374 |
Gene Symbol: LOC102551046 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770812.1 | XP_008769034.1 | 5 | 936 |