Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014239.1 | NP_001014261.1 | 9 | 1648 | ||
GenBank mRNA | BC082088.1 | - | 9 | 1648 | 7133991 |