Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014244.1 | NP_001014266.1 | 3, 4 | 329 | ||
XM_008759774.1 | XP_008757996.1 | 1, 2 | 1467 | |||
XM_008759775.1 | XP_008757997.1 | 2, 3 | 1370 | |||
XM_008759776.1 | XP_008757998.1 | 1, 2 | 1467 | |||
XM_008759777.1 | XP_008757999.1 | 1, 2 | 1467 | |||
XM_008759778.1 | XP_008758000.1 | 2, 3 | 785 | |||
XM_008759779.1 | XP_008758001.1 | 2, 3 | 942 | |||
XM_008759781.1 | XP_008758003.1 | 3, 4 | 266 | |||
XM_008759782.1 | XP_008758004.1 | 1, 2 | 1467 | |||
GenBank mRNA | BC079235.1 | - | 3, 4 | 329 | 7132617 |