Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lzts2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014247.2 | NP_001014269.2 | 4 | 2485 | ||
XM_006231547.2 | XP_006231609.1 | 5 | 3535 | |||
XM_006231548.2 | XP_006231610.1 | 6 | 2736 | |||
XM_006231549.2 | XP_006231611.1 | 5 | 3964 | |||
XM_008760444.1 | XP_008758666.1 | 6 | 2727 | |||
GenBank mRNA | BC083580.1 | - | 7 | 2291 | 7129196 |