Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017864.3 | NP_060334.2 | 15 | 1977 | ||
NR_073444.1 | 15 | 1977 | ||||
NR_073445.1 | 15 | 1946 | ||||
XM_017013616.1 | XP_016869105.1 | 15 | 1977 | |||
XM_017013617.1 | XP_016869106.1 | 15 | 1977 | |||
XM_017013618.1 | XP_016869107.1 | 12 | 1530 | |||
XM_017013619.1 | XP_016869108.1 | 15 | 1936 | |||
GenBank mRNA | AB161944.1 | - | 15 | 1961 | ||
AK000537.1 | - | 5 | 531 | |||
AK001403.1 | - | 8 | 917 | |||
AK091278.1 | - | 14 | 1714 | |||
AK225533.1 | - | 5 | 531 | |||
BC050536.1 | - | 7 | 765 | |||
BC064915.1 | - | 7 | 806 | |||
BC136754.1 | - | 15 | 1891 | 9020744 | ||
BC144396.1 | - | 15 | 1891 | |||
BC144397.1 | - | 15 | 1860 | |||
BX538203.1 | - | 11 | 1300 |