Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAXX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141969.1 | NP_001135441.1 | 3 | 464 | ||
NM_001141970.1 | NP_001135442.1 | 3 | 395 | |||
NM_001254717.1 | NP_001241646.1 | 2 | 205 | |||
NM_001350.4 | NP_001341.1 | 3 | 461 | |||
GenBank mRNA | AB209493.1 | - | 1 | 630 | ||
AF015956.2 | - | 2 | 311 | |||
AF039136.1 | - | 3 | 404 | |||
AF050179.1 | - | 3 | 372 | |||
AF097742.1 | - | 2 | 264 | |||
AK223404.1 | - | 3 | 462 | |||
AK292187.1 | - | 3 | 462 | |||
AK303767.1 | - | 3 | 338 | |||
AK303854.1 | - | 2 | 162 | |||
AK309612.1 | - | 2 | 514 | |||
AK313034.1 | - | 3 | 404 | |||
BC109073.1 | - | 3 | 347 | 40034214 | ||
BC109074.1 | - | 3 | 347 | 40034219 | ||
CR457085.1 | - | 2 | 257 | |||
DC345308.1 | - | 3 | 461 | |||
HQ436528.1 | - | 2 | 257 | |||
HQ436529.1 | - | 2 | 257 |