Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014254.1 | NP_001014276.1 | 2, 3 | 444 | ||
XM_008761188.1 | XP_008759410.1 | 2, 3 | 628 | |||
XM_008761189.1 | XP_008759411.1 | 2, 3 | 628 | |||
GenBank mRNA | BC083788.1 | - | 2, 3 | 444 | 7098251 |