Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017867.2 | NP_060337.2 | 3 | 393 | ||
XM_017008332.1 | XP_016863821.1 | 3 | 405 | |||
GenBank mRNA | AK000541.1 | - | 3 | 348 | ||
AL136673.1 | - | 3 | 363 | |||
BC010367.1 | - | 3 | 359 | 4334172 | ||
BM905677.1 | - | 3 | 313 | |||
CB141427.1 | - | 3 | 393 | |||
DN990093.1 | - | 3 | 360 |