Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tceanc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014260.2 | NP_001014282.1 | 5 | 1049 | ||
GenBank mRNA | BC083757.1 | - | 5 | 1036 | 7192791 |