Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Azin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC078981.1 | - | 181 | 7112100 | ||
CK470797.1 | - | 385 | ||||
HQ413774.1 | - | 1 | 30 |