Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptges3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014272.1 | NP_001014294.1 | 2 | 1122 | ||
GenBank mRNA | BC087125.1 | - | 2 | 1122 | 7110309 |