Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF407 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146189.1 | NP_001139661.1 | 3, 4 | 4942 | ||
NM_017757.2 | NP_060227.2 | 3, 4 | 4942 | |||
XM_005266726.4 | XP_005266783.1 | 4, 5 | 5141 | |||
XM_011526068.2 | XP_011524370.1 | 4, 5 | 8648 | |||
XM_017025838.1 | XP_016881327.1 | 4, 5 | 5006 | |||
GenBank mRNA | AB051490.1 | - | 3, 4 | 1638 | ||
AK000314.1 | - | 3, 4 | 2567 |