Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNASE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001375.2 | NP_001366.1 | 3 | 466 | ||
GenBank mRNA | AB004574.1 | - | 3 | 392 | ||
AF045937.1 | - | 3 | 396 | |||
AF047016.1 | - | 3 | 392 | |||
AF060222.1 | - | 3 | 413 | |||
AK297493.1 | - | 3 | 394 | |||
AK311145.1 | - | 3 | 415 | |||
AK315358.1 | - | 3 | 408 | |||
BC010419.2 | - | 3 | 344 | 3528108 | ||
BC065209.1 | - | 3 | 411 | 5502061 | ||
BT007047.1 | - | 3 | 320 | |||
BX394955.2 | - | 3 | 516 | |||
FJ752529.1 | - | 3 | 272 |