Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015029.2 | NP_001015029.2 | 1 | 87 | ||
GenBank mRNA | AY779030.1 | - | 1 | 37 | ||
BC105813.1 | - | 1 | 87 | 7112648 |