Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3gntl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015035.1 | NP_001015035.1 | 6 | 576 | ||
XM_006247904.2 | XP_006247966.1 | 6 | 626 | |||
XM_008768503.1 | XP_008766725.1 | 6 | 626 | |||
XM_008768504.1 | XP_008766726.1 | 6 | 626 | |||
XM_008768505.1 | XP_008766727.1 | 6 | 626 | |||
XM_008768506.1 | XP_008766728.1 | 6 | 626 | |||
XM_008768507.1 | XP_008766729.1 | 6 | 626 | |||
XM_008768508.1 | XP_008766730.1 | 6 | 626 | |||
XM_008768509.1 | XP_008766731.1 | 6 | 626 | |||
XM_008768510.1 | XP_008766732.1 | 4 | 329 | |||
GenBank mRNA | AY634365.1 | - | 6 | 576 | ||
BC111711.1 | - | 6 | 550 | 7383845 |