Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC361914 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017465.1 | NP_001017465.1 | 1 | 731 | ||
XM_006232112.2 | XP_006232174.1 | 2 | 665 | |||
GenBank mRNA | BC079349.1 | - | 1 | 731 | 7107785 |
Gene Symbol: LOC100910566 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232118.2 | XP_006232180.1 | 2 | 665 |