Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017785.4 | NP_060255.3 | 4 | 701 | ||
XM_011534585.1 | XP_011532887.1 | 4 | 647 | |||
XM_011534586.2 | XP_011532888.1 | 4 | 583 | |||
XM_011534587.2 | XP_011532889.1 | 5 | 848 | |||
XM_011534588.2 | XP_011532890.1 | 5 | 786 | |||
XM_011534589.2 | XP_011532891.1 | 4 | 596 | |||
XM_011534590.2 | XP_011532892.1 | 3 | 337 | |||
XM_017009601.1 | XP_016865090.1 | 4 | 532 | |||
XM_017009602.1 | XP_016865091.1 | 3 | 337 | |||
GenBank mRNA | AF269167.1 | - | 4 | 592 | ||
AK000371.1 | - | 3 | 427 | |||
AK074227.1 | - | 1 | 106 | |||
AK308835.1 | - | 4 | 580 | |||
AK310442.1 | - | 4 | 500 | |||
AY517557.1 | - | 4 | 532 | |||
BC012568.1 | - | 4 | 567 | 4102558 |