Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zw10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024801.1 | NP_001019972.1 | 9 | 1282 | ||
GenBank mRNA | BC097452.1 | - | 9 | 1282 | 7456845 |