Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024898.1 | NP_001020069.1 | 5 | 516 | ||
GenBank mRNA | BC097494.1 | - | 5 | 516 | 7461781 |