Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024902.1 | NP_001020073.1 | 4, 5 | 433 | ||
XM_006230564.2 | XP_006230626.1 | 4, 5 | 919 | |||
XM_006230565.2 | XP_006230627.1 | 4, 5 | 1029 | |||
XM_006230566.2 | XP_006230628.1 | 4, 5 | 1202 | |||
XM_006230569.2 | XP_006230631.1 | 4, 5 | 956 | |||
XM_006230570.2 | XP_006230632.1 | 2, 3 | 166 | |||
XM_008760005.1 | XP_008758227.1 | 4, 5 | 1201 | |||
GenBank mRNA | BC097444.1 | - | 4, 5 | 433 | 7456428 |