Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017793.2 | NP_060263.2 | 1 | 2006 | ||
GenBank mRNA | AK000381.1 | - | 2 | 1880 | ||
AY034074.1 | - | 2 | 1060 | |||
BC002497.2 | - | 1 | 1324 | 3050784 | ||
BC007270.1 | - | 1 | 1336 | 3160319 | ||
BE254727.1 | - | 1 | 261 |