Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025023.1 | NP_001020194.1 | 18 | 2324 | ||
GenBank mRNA | BC079443.1 | - | 18 | 2324 | 7115485 |