Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bag4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025130.1 | NP_001020301.1 | 3 | 1033 | ||
GenBank mRNA | BC083608.1 | - | 3 | 1033 | 7133487 |