Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnih2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025132.1 | NP_001020303.1 | 6 | 726 | ||
XM_008760153.1 | XP_008758375.1 | 5 | 1304 | |||
XM_008760154.1 | XP_008758376.1 | 6 | 1364 | |||
GenBank mRNA | BC091325.1 | - | 6 | 726 | 7313787 |