Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LFNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002304.2 | NP_002295.1 | 1, 2 | 86 | ||
GenBank mRNA | BC014851.1 | - | 1, 2 | 86 | 4453156 |