Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005716.3 | NP_005707.1 | 1938 | |||
NM_202468.2 | NP_974197.1 | 1882 | ||||
NM_202469.2 | NP_974198.1 | 1564 | ||||
NM_202470.2 | NP_974199.1 | 1794 | ||||
NM_202494.2 | NP_974223.1 | 1476 |