Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eef1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010106.2 | NP_034236.2 | 6 | 1112 | ||
GenBank mRNA | AK003188.1 | - | 6 | 1049 | ||
AK011771.1 | - | 6 | 1061 | |||
AK012341.1 | - | 6 | 1061 | |||
AK032914.1 | - | 6 | 1058 | |||
AK050635.1 | - | 6 | 1061 | |||
AK076696.1 | - | 6 | 1060 | |||
AK081725.1 | - | 6 | 1060 | |||
AK083361.1 | - | 6 | 1060 | |||
AK133725.1 | - | 6 | 1060 | |||
AK135448.1 | - | 6 | 1061 | |||
AK137223.1 | - | 6 | 1060 | |||
AK141243.1 | - | 6 | 1062 | |||
AK150798.1 | - | 6 | 1060 | |||
AK151479.1 | - | 6 | 1061 | |||
AK151773.1 | - | 6 | 1060 | |||
AK152810.1 | - | 6 | 1060 | |||
AK152993.1 | - | 6 | 1061 | |||
AK153387.1 | - | 6 | 1060 | |||
AK153402.1 | - | 6 | 1060 | |||
AK153851.1 | - | 6 | 1062 | |||
AK162049.1 | - | 6 | 1060 | |||
AK162313.1 | - | 6 | 1059 | |||
AK162440.1 | - | 6 | 1064 | |||
AK163799.1 | - | 6 | 1060 | |||
AK164113.1 | - | 6 | 1062 | |||
AK164671.1 | - | 6 | 1060 | |||
AK165938.1 | - | 6 | 1060 | |||
AK166021.1 | - | 6 | 1060 | |||
AK166033.1 | - | 6 | 1060 | |||
AK166058.1 | - | 6 | 1060 | |||
AK166089.1 | - | 6 | 1060 | |||
AK166615.1 | - | 6 | 1060 | |||
AK166807.1 | - | 6 | 1060 | |||
AK167843.1 | - | 6 | 1060 | |||
AK167873.1 | - | 6 | 1059 | |||
AK168424.1 | - | 6 | 1062 | |||
AK168467.1 | - | 6 | 1068 | |||
AK183068.1 | - | 1 | 51 | |||
AK185520.1 | - | 1 | 51 | |||
AK194927.1 | - | 1 | 51 | |||
AK208452.1 | - | 1 | 51 | |||
BC003969.1 | - | 5 | 1030 | |||
BC004005.1 | - | 5 | 1021 | 3492770 | ||
BC004067.1 | - | 5 | 1049 | 3590965 | ||
BC005660.1 | - | 5 | 1050 | 3588966 | ||
BC018223.1 | - | 6 | 1048 | 4012653 | ||
BC018485.1 | - | 6 | 1062 | 3491827 | ||
BC040689.1 | - | 3 | 552 | |||
BC050124.1 | - | 2605 | ||||
BC068258.1 | - | 2857 | ||||
BC083069.1 | - | 6 | 1048 | 6310448 | ||
BC092053.1 | - | 5 | 1038 | 4008574 | ||
BC092276.1 | - | 5 | 1043 | 3490007 | ||
BC092292.1 | - | 5 | 1003 | |||
BC095965.1 | - | 5 | 1038 | 6310404 | ||
BC108391.1 | - | 6 | 1045 | 30242277 | ||
M22432.1 | - | 6 | 1051 | |||
X13661.1 | - | 5 | 1028 |
Gene Symbol: Gm6548 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC089461.1 | - | 1 | 1358 |