Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027976.2 | NP_082252.1 | 21 | 2877 | ||
GenBank mRNA | AK006541.1 | - | 10 | 1354 | ||
AK034942.1 | - | 2877 | ||||
AK145894.1 | - | 21 | 2538 | |||
AK150542.1 | - | 21 | 2543 | |||
AK150663.1 | - | 21 | 2543 | |||
AK151378.1 | - | 21 | 2332 | |||
AK161024.1 | - | 21 | 2230 | |||
AK168019.1 | - | 9 | 878 | |||
AK182997.1 | - | 2 | 166 | |||
AK214922.1 | - | 1 | 135 | |||
BC031544.1 | - | 21 | 2311 | 3987201 |