Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019503.4 | NP_062376.2 | 3 | 193 | ||
NM_052991.4 | NP_443717.1 | 3 | 176 | |||
NM_052992.3 | NP_443718.1 | 3 | 257 | |||
NM_194321.2 | NP_919302.1 | 3 | 240 | |||
GenBank mRNA | AA462069.1 | - | 3 | 125 | ||
AA466595.1 | - | 3 | 193 | |||
AF089734.1 | - | 3 | 119 | |||
AK002585.1 | - | 3 | 180 | |||
BC024671.1 | - | 3 | 130 | 4208962 | ||
BY064152.1 | - | 3 | 192 | |||
BY132051.1 | - | 3 | 162 | |||
CA459152.1 | - | 3 | 198 | |||
CK793316.1 | - | 3 | 175 | |||
W16239.1 | - | 3 | 156 | |||
W98524.1 | - | 3 | 171 |