Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Artn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053397.1 | NP_445849.1 | 5 | 1218 | ||
XM_006238684.2 | XP_006238746.1 | 3 | 4732 | |||
XM_006238685.2 | XP_006238747.1 | 4 | 4513 | |||
XM_006238687.2 | XP_006238749.1 | 4 | 4468 | |||
XM_006238691.2 | XP_006238753.1 | 3 | 4736 | |||
GenBank mRNA | BC079078.1 | - | 5 | 1218 | 7122331 |