Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005554.3 | NP_005545.1 | 9 | 2030 | ||
GenBank mRNA | BC008807.2 | - | 9 | 1869 | 3947610 | |
BC139753.1 | - | 9 | 1822 | 40117016 |
Gene Symbol: KRT6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173086.4 | NP_775109.2 | 9 | 1869 | ||
GenBank mRNA | AK299478.1 | - | 10 | 1873 | ||
BC110639.1 | - | 9 | 1869 | 4754834 | ||
L42611.1 | - | 9 | 1861 |