Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm10408 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256501.1 | NP_001243430.1 | 5 | 1218 |
Gene Symbol: D830030K20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177135.4 | NP_796109.1 | 5 | 1076 | ||
GenBank mRNA | AK085928.1 | - | 5 | 1077 |
Gene Symbol: Gm3715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017182.1 | - | 772 |