Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204883.1 | NP_001191812.1 | 4 | 1784 | ||
NM_001204884.1 | NP_001191813.1 | 4 | 1772 | |||
NM_001204885.1 | NP_001191814.1 | 4 | 1762 | |||
NM_001204886.1 | NP_001191815.1 | 4 | 1799 | |||
NM_001204887.1 | NP_001191816.1 | 4 | 2144 | |||
NM_001204888.1 | NP_001191817.1 | 3 | 2036 | |||
NM_198490.2 | NP_940892.1 | 3 | 2050 | |||
GenBank mRNA | AK128345.1 | - | 3 | 1846 |
Gene Symbol: ISY1-RAB43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204890.1 | NP_001191819.1 | 13 | 2637 | ||
GenBank mRNA | AB032986.1 | - | 13 | 2443 |