Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn2r113 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104578.1 | NP_001098048.1 | 4 | 1330 | ||
GenBank mRNA | AB540949.1 | - | 4 | 1330 |
Gene Symbol: Vmn2r39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105071.1 | NP_001098541.1 | 4 | 1333 |
Gene Symbol: Vmn2r43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198961.2 | NP_945199.1 | 4 | 1325 | ||
GenBank mRNA | AJ543405.1 | - | 4 | 1327 |
Set Membership: |
G-protein coupled receptor |