Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201423.2 | NP_958826.1 | 1 | 371 | ||
GenBank mRNA | AY435130.1 | - | 1 | 371 | ||
M34007.1 | - | 1 | 365 |