Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202554.1 | NP_001189483.1 | 3 | 1776 | ||
NM_017837.3 | NP_060307.2 | 3 | 1450 | |||
GenBank mRNA | AK000484.1 | - | 3 | 1225 | ||
AK056539.1 | - | 3 | 1769 | |||
AK225521.1 | - | 3 | 1225 | |||
AK301626.1 | - | 4 | 755 | |||
BC013568.1 | - | 3 | 1418 | 3890907 |