Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164246.1 | NP_001157718.1 | 8 | 875 | ||
NM_017847.5 | NP_060317.3 | 9 | 971 | |||
XM_011509660.1 | XP_011507962.1 | 9 | 970 | |||
XM_011509661.1 | XP_011507963.1 | 9 | 970 | |||
XM_017001543.1 | XP_016857032.1 | 9 | 970 | |||
GenBank mRNA | AK000512.1 | - | 9 | 891 | ||
AK026086.1 | - | 7 | 541 | |||
AK298107.1 | - | 8 | 811 | |||
AY854248.1 | - | 9 | 864 | |||
BC003397.1 | - | 9 | 804 | 3448518 | ||
DQ323046.1 | - | 8 | 735 |