Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017855.3 | NP_060325.3 | 9 | 680 | ||
XM_005265698.3 | XP_005265755.1 | 10 | 744 | |||
XM_006714245.3 | XP_006714308.1 | 9 | 673 | |||
XM_017008331.1 | XP_016863820.1 | 8 | 636 | |||
GenBank mRNA | AK098169.1 | - | 2 | 374 | ||
BC017796.1 | - | 5 | 287 | 4689080 | ||
EF113908.1 | - | 9 | 682 |