Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136036.2 | NP_001129508.1 | 8 | 1362 | ||
NM_001193328.1 | NP_001180257.1 | 7 | 914 | |||
NM_017865.3 | NP_060335.2 | 8 | 1146 | |||
XM_011544220.1 | XP_011542522.1 | 8 | 1403 | |||
XM_011544221.1 | XP_011542523.1 | 8 | 1400 | |||
XM_011544222.1 | XP_011542524.1 | 8 | 1018 | |||
XM_011544223.2 | XP_011542525.1 | 7 | 1268 | |||
XM_011544227.1 | XP_011542529.1 | 8 | 1314 | |||
XM_017001698.1 | XP_016857187.1 | 8 | 1015 | |||
XM_017001701.1 | XP_016857190.1 | 8 | 1130 | |||
XM_017001703.1 | XP_016857192.1 | 8 | 1333 | |||
XM_017001704.1 | XP_016857193.1 | 8 | 1290 | |||
XM_017001705.1 | XP_016857194.1 | 8 | 1193 | |||
GenBank mRNA | AB209706.1 | - | 4 | 1200 | ||
AF256175.1 | - | 1 | 463 | |||
AK000538.1 | - | 7 | 883 | |||
AK001070.1 | - | 6 | 562 | |||
AK225535.1 | - | 7 | 883 | |||
AK298899.1 | - | 6 | 1091 | |||
AK302187.1 | - | 8 | 1296 | |||
AL133100.1 | - | 6 | 1875 | |||
BC002948.1 | - | 8 | 1102 | 3533691 |