Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sumo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024295.1 | NP_001019466.1 | 4 | 531 | ||
GenBank mRNA | BC083728.1 | - | 4 | 531 | 7191303 |