Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024304.1 | NP_001019475.1 | 8 | 1473 | ||
GenBank mRNA | BC091199.1 | - | 8 | 1473 | 7379741 | |
FQ223925.1 | - | 7 | 1335 |