Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hyal6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024320.1 | NP_001019491.1 | 4 | 2032 | ||
XM_006236165.2 | XP_006236227.1 | 3 | 1954 | |||
GenBank mRNA | BC082016.1 | - | 4 | 2032 | 7132110 |