Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024322.1 | NP_001019493.1 | 4 | 319 | ||
XM_006236160.2 | XP_006236222.1 | 5 | 411 | |||
XM_006236161.2 | XP_006236223.1 | 6 | 633 | |||
XM_008762809.1 | XP_008761031.1 | 6 | 815 | |||
XM_008762810.1 | XP_008761032.1 | 6 | 760 | |||
XM_008762811.1 | XP_008761033.1 | 4 | 364 | |||
GenBank mRNA | BC085834.1 | - | 4 | 319 | 7134840 |