Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC500077 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024325.1 | NP_001019496.1 | 4 | 870 | ||
XM_006236275.2 | XP_006236337.1 | 3 | 1747 | |||
GenBank mRNA | BC079043.1 | - | 4 | 870 | 7114933 |