Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp467 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024327.1 | NP_001019498.1 | 4 | 2303 | ||
XM_006236431.2 | XP_006236493.1 | 5 | 2388 | |||
XM_006236432.2 | XP_006236494.1 | 5 | 2638 | |||
XM_006236433.2 | XP_006236495.1 | 5 | 2919 | |||
XM_006236434.2 | XP_006236496.1 | 5 | 2496 | |||
GenBank mRNA | BC086534.1 | - | 4 | 2303 | 7097755 |