Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC500331 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024337.1 | NP_001019508.1 | 5 | 932 | ||
XM_006237415.2 | XP_006237477.1 | 4 | 631 | |||
XM_008763321.1 | XP_008761543.1 | 3 | 823 | |||
XM_008763322.1 | XP_008761544.1 | 4 | 719 | |||
GenBank mRNA | BC086997.1 | - | 5 | 932 | 7131139 |
Gene Symbol: Clec2dl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085404.1 | NP_001078873.1 | 3 | 394 | ||
XM_006237453.2 | XP_006237515.1 | 4 | 427 | |||
XM_008763333.1 | XP_008761555.1 | 3 | 1453 | |||
GenBank mRNA | EF100689.1 | - | 3 | 394 |
Gene Symbol: LOC100910725 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237437.2 | XP_006237499.1 | 4 | 503 | ||
XM_006237438.2 | XP_006237500.1 | 418 | ||||
XM_008763340.1 | XP_008761562.1 | 823 |