Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024349.2 | NP_001019520.2 | 2 | 218 | ||
XM_008764373.1 | XP_008762595.1 | 2 | 1473 | |||
XM_008764374.1 | XP_008762596.1 | 3 | 1391 | |||
GenBank mRNA | BC088265.1 | - | 2 | 219 | 7377562 |