Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cesl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024365.1 | NP_001019536.1 | 4, 5 | 522 | ||
GenBank mRNA | AB023630.1 | - | 437 | |||
BC079129.1 | - | 4, 5 | 522 | 7127228 | ||
U10698.1 | - | 3, 4 | 409 |
Gene Symbol: Ces1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772260.1 | XP_008770482.1 | 4, 5 | 563 | ||
XM_008772261.1 | XP_008770483.1 | 4, 5 | 533 | |||
XM_008772262.1 | XP_008770484.1 | 3, 4 | 466 |