Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024909.1 | NP_001020080.1 | 1 | 1166 | ||
GenBank mRNA | BC097418.1 | - | 1 | 1166 | 7454547 |