Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ier5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025041.1 | NP_001020212.1 | 1 | 1122 | ||
GenBank mRNA | BC087095.1 | - | 1 | 1122 | 7191438 |